AC5372909657 Accela ChemBio Inc.

5-Hydroxy-7-methyl-1H-indazole

  • MDL number MFCD12198892
  • Cas Number : 478841-61-1
  • Molecular Weight : 148.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY005855&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY005855.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart