AC5372909682 Accela ChemBio Inc.

5-Bromo-2-methyl-3-nitrobenzoic Acid

  • MDL number MFCD07357316
  • Cas Number : 107650-20-4
  • Molecular Weight : 260.0415

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY005907&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY005907.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart