AC5372909701 Accela ChemBio Inc.

Methyl 3-(Morpholinomethyl)benzoate

  • MDL number MFCD07772832
  • Cas Number : 190660-95-8
  • Molecular Weight : 235.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY005959&attname=GoodsCode

Safety Information

signalword Danger
pcodes P301 + P330 + P331-P280-P305 + P351 + P338-P310- P301+ P312
hcodes GHS05, GHS07
MSDS http://pdf.accelachem.com/cn/msds/SY005959.pdf
hstatements H302-H318-H314
Availability details for
Quantity
  • Availability
  • Add to Cart