AC5372909742 Accela ChemBio Inc.

2-Fluoro-4-(methylsulfonyl)nitrobenzene

  • MDL number MFCD12828682
  • Cas Number : 1166756-97-3
  • Molecular Weight : 219.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY006066&attname=GoodsCode

Safety Information

hcodes No data known
MSDS http://pdf.accelachem.com/cn/msds/SY006066.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart