AC5372909747 Accela ChemBio Inc.

2-Isopropoxyaniline

  • MDL number MFCD00242947
  • Cas Number : 29026-74-2
  • Molecular Weight : 151.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY006071&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY006071.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart