AC5372909748 Accela ChemBio Inc.

6-Aminoquinazoline

  • MDL number MFCD10697883
  • Cas Number : 101421-72-1
  • Molecular Weight : 145.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY006072&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY006072.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart