AC5372909767 Accela ChemBio Inc.

Ethyl 3-Oxopiperidine-4-carboxylate Hydrochloride

  • MDL number MFCD12828687
  • Cas Number : 72738-09-1
  • Molecular Weight : 207.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY006124&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY006124.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart