AC5372909786 Accela ChemBio Inc.

6-Acetylquinoline

  • MDL number MFCD00047614
  • Cas Number : 73013-68-0
  • Molecular Weight : 171.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form White to brown solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY006186&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY006186.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart