AC5372909808 Accela ChemBio Inc.

3-Bromoindole-5-carboxylic Acid

  • MDL number MFCD12828698
  • Cas Number : 916179-87-8
  • Molecular Weight : 240.05344

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20?
Physical Form solid
Shipping Temperature +4 ?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY006247&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY006247.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart