AC5372909838 Accela ChemBio Inc.

2-(Methylsulfonyl)-4-pyridinecarboxylic Acid

  • MDL number MFCD12828707
  • Cas Number : 1186663-27-3
  • Molecular Weight : 201.19978

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY006343&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY006343.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart