AC5372909840 Accela ChemBio Inc.

2-Methyl-3-(methylsulfonyl)benzoic Acid

  • MDL number MFCD12828710
  • Cas Number : 1186663-49-9
  • Molecular Weight : 214.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY006348&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY006348.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart