AC5372909875 Accela ChemBio Inc.

N-Boc-DL-phenylalaninol

  • MDL number MFCD01457313
  • Cas Number : 145149-48-0
  • Molecular Weight : 251.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY006461&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY006461.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart