AC5372909895 Accela ChemBio Inc.

4-Aminopyrimido[4,5-d]pyrimidine

  • MDL number MFCD02251673
  • Cas Number : 26979-05-5
  • Molecular Weight : 147.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY006526&attname=GoodsCode

Safety Information

hcodes No data known
MSDS http://pdf.accelachem.com/cn/msds/SY006526.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart