AC5372909898 Accela ChemBio Inc.

4-(4-Ethyl-1-piperazinyl)-2-fluoroaniline Hydrochloride

  • MDL number MFCD13191588
  • Cas Number : 1197193-28-4
  • Molecular Weight : 259.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form White to tan solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY006646&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY006646.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart