AC5372909915 Accela ChemBio Inc.

2-(Cyclopentyloxy)acetic Acid

  • MDL number MFCD09930082
  • Cas Number : 95832-60-3
  • Molecular Weight : 144.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY006722&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY006722.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart