AC5372909937 Accela ChemBio Inc.

5,6,7,8-Tetrahydro-1,2,4-triazolo[4,3-a]pyrazine Hydrochloride

  • MDL number MFCD08448159
  • Cas Number : 837430-14-5
  • Molecular Weight : 160.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form Yellow to brown solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY006775&attname=GoodsCode

Safety Information

signalword Danger
pcodes P301+P310
MSDS http://pdf.accelachem.com/cn/msds/SY006775.pdf
hstatements H301
Availability details for
Quantity
  • Availability
  • Add to Cart