AC5372909956 Accela ChemBio Inc.

4-(4-Methyl-1-piperazinyl)-2-(methylsulfonyl)benzaldehyde

  • MDL number MFCD13191596
  • Cas Number : 1197193-26-2
  • Molecular Weight : 282.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY006890&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY006890.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart