AC5372909957 Accela ChemBio Inc.

2-Fluoro-4-(4-methyl-1-piperazinyl)benzaldehyde

  • MDL number MFCD13191597
  • Cas Number : 1197193-42-2
  • Molecular Weight : 222.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY006891&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY006891.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart