AC5372909960 Accela ChemBio Inc.

4-Fluoro-3-pyrrolidinobenzaldehyde

  • MDL number MFCD13191603
  • Cas Number : 1197193-31-9
  • Molecular Weight : 193.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20?
Physical Form Light yellow to purple to brown liquid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY006898&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY006898.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart