AC5372909964 Accela ChemBio Inc.

5-Fluoro-2-(4-methyl-1-piperazinyl)benzoic Acid

  • MDL number MFCD11639778
  • Cas Number : 1096829-46-7
  • Molecular Weight : 238.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY006906&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY006906.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart