AC5372909966 Accela ChemBio Inc.

2-Fluoro-4-morpholinobenzoic Acid

  • MDL number MFCD11040390
  • Cas Number : 946598-40-9
  • Molecular Weight : 225.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY006917&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY006917.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart