AC5372909972 Accela ChemBio Inc.

3-Fluoro-4-morpholinobenzoic Acid

  • MDL number MFCD11156507
  • Cas Number : 588708-72-9
  • Molecular Weight : 225.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY006934&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY006934.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart