AC5372909975 Accela ChemBio Inc.

3-Chloro-4-(methylsulfonyl)benzoic Acid

  • MDL number MFCD13191622
  • Cas Number : 1197193-45-5
  • Molecular Weight : 234.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form White to Off-white solid or powder
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY006942&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY006942.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart