AC5372909979 Accela ChemBio Inc.

3-Chloro-4-(4-methyl-1-piperazinyl)benzoic Acid

  • MDL number MFCD13191625
  • Cas Number : 1197193-33-1
  • Molecular Weight : 254.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY006946&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY006946.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart