AC5372910014 Accela ChemBio Inc.

Diethyl 2-Chloro-4,5-pyrimidinedicarboxylate

  • MDL number MFCD16659907
  • Cas Number : 90794-84-6
  • Molecular Weight : 258.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form White to brown solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY007070&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY007070.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart