AC5372910042 Accela ChemBio Inc.

Methyl 6-Methylpyridazine-3-carboxylate

  • MDL number MFCD13174778
  • Cas Number : 106584-51-4
  • Molecular Weight : 152.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY007132&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY007132.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart