AC5372910066 Accela ChemBio Inc.

2-(Hydroxymethyl)homomorpholine Hydrochloride

  • MDL number MFCD14585014
  • Cas Number : 1207194-51-1
  • Molecular Weight : 167.63386

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY007204&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY007204.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart