AC5372910089 Accela ChemBio Inc.

2,4-Dichloro-8-fluoroquinazoline

  • MDL number MFCD09954886
  • Cas Number : 959237-64-0
  • Molecular Weight : 217.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY007267&attname=GoodsCode

Safety Information

hcodes No data known
MSDS http://pdf.accelachem.com/cn/msds/SY007267.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart