AC5372910173 Accela ChemBio Inc.

5-Amino-4,6-dihydroxypyrimidine

  • MDL number MFCD00239457
  • Cas Number : 69340-97-2
  • Molecular Weight : 127.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY007614&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY007614.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart