AC5372910208 Accela ChemBio Inc.

Diethyl Pyrazolo[1,5-a]pyridine-2,3-dicarboxylate

  • MDL number MFCD16495867
  • Cas Number : 1226776-92-6
  • Molecular Weight : 262.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form Red to dark brown liquid or low melting solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY007786&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY007786.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart