AC5372910253 Accela ChemBio Inc.

Methyl 4-(4-Methylpiperazinomethyl)benzoate

  • MDL number MFCD01467150
  • Cas Number : 314268-40-1
  • Molecular Weight : 248.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY008016&attname=GoodsCode

Safety Information

signalword Danger
pcodes P301+P330+P331-P280-P305+P351+P338-P310-P301+P312
hcodes GHS07
MSDS http://pdf.accelachem.com/cn/msds/SY008016.pdf
hstatements H314-H302-H318
Availability details for
Quantity
  • Availability
  • Add to Cart