AC5372910284 Accela ChemBio Inc.

4-Methoxy-2,6-dimethylbenzenesulfonyl Chloride

  • MDL number MFCD00210327
  • Cas Number : 55661-08-0
  • Molecular Weight : 234.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20?
Physical Form liquid
Shipping Temperature +4 ?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY008164&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY008164.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart