AC5372910296 Accela ChemBio Inc.

(R)-1-(3-Pyridyl)ethanol

  • MDL number MFCD07368883
  • Cas Number : 7606-26-0
  • Molecular Weight : 123.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form Colorless to yellow liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY008212&attname=GoodsCode

Safety Information

signalword Warning
hcodes GHS07
MSDS http://pdf.accelachem.com/cn/msds/SY008212.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart