AC5372910341 Accela ChemBio Inc.

(3-Cyanophenoxy)acetic Acid

  • MDL number MFCD00516888
  • Cas Number : 1879-58-9
  • Molecular Weight : 177.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form White to off white solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY008339&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY008339.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart