AC5372910350 Accela ChemBio Inc.

1-Cyclopropyl-1,3-dihydroimidazo[4,5-c]pyridine-2-one

  • MDL number MFCD09954763
  • Cas Number : 380605-29-8
  • Molecular Weight : 175.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY008382&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY008382.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart