AC5372910357 Accela ChemBio Inc.

7-Boc-3-phenyl-5,6,7,8-tetrahydroimidazo[1,2-a]pyrazine

  • MDL number MFCD16495904
  • Cas Number : 1226776-87-9
  • Molecular Weight : 299.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY008393&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY008393.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart