AC5372910360 Accela ChemBio Inc.

1-Benzyl-4-hydroxypiperidine-4-carboxylic Acid

  • MDL number MFCD06208305
  • Cas Number : 59119-18-5
  • Molecular Weight : 235.27898

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY008401&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY008401.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart