AC5372910389 Accela ChemBio Inc.

N-[4-(2-tert-Butylphenylsulfonyl)phenyl]-2,3,4-trihydroxy-5-(2-isopropylbenzyl)benzamide

  • MDL number MFCD17010275
  • Cas Number : 877877-35-5
  • Molecular Weight : 573.6991

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form Purplish red solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY008562&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY008562.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart