AC5372910394 Accela ChemBio Inc.

(R)-2-(3-Fluorophenyl)pyrrolidine Hydrochloride

  • MDL number MFCD08751398
  • Cas Number : 1364890-61-8
  • Molecular Weight : 201.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY008576&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY008576.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart