AC5372910395 Accela ChemBio Inc.

Ethyl 2-[1-(6-Chloro-3-pyridazinyl)-4-piperidyl]acetate

  • MDL number MFCD17171371
  • Cas Number : 1246471-48-6
  • Molecular Weight : 283.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form Light yellow to brown powder or solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY008582&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY008582.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart