AC5372910396 Accela ChemBio Inc.

Methyl 1-(6-Chloro-3-pyridazinyl)piperidine-3-carboxylate

  • MDL number MFCD07368264
  • Cas Number : 1208086-32-1
  • Molecular Weight : 255.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form Yellow to orange solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY008583&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY008583.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart