AC5372910397 Accela ChemBio Inc.

3-Chloro-6-(4-piperidyloxy)pyridazine

  • MDL number MFCD16713758
  • Cas Number : 1185536-63-3
  • Molecular Weight : 213.66408

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY008586&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY008586.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart