AC5372910403 Accela ChemBio Inc.

4-(5-Nitrobenzimidazol-2-yl)aniline

  • MDL number MFCD00430514
  • Cas Number : 71002-88-5
  • Molecular Weight : 254.2441

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY008602&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY008602.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart