AC5372910405 Accela ChemBio Inc.

3-(5-Methylisoxazol-3-yl)-6-[(1-methyl-1H-1,2,3-triazol-4-yl)methoxy][1,2,4]triazolo[3,4-a]phthalazine

  • MDL number MFCD09832717
  • Cas Number : 215874-86-5
  • Molecular Weight : 362.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY008621&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY008621.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart