AC5372910408 Accela ChemBio Inc.

3-Ethynylindole

  • MDL number MFCD00463781
  • Cas Number : 62365-78-0
  • Molecular Weight : 141.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY008634&attname=GoodsCode

Safety Information

signalword Danger
pcodes P280;P305+P351+P338
hcodes GHS07
MSDS http://pdf.accelachem.com/cn/msds/SY008634.pdf
hstatements H301
Availability details for
Quantity
  • Availability
  • Add to Cart