AC5372910410 Accela ChemBio Inc.

Methyl 2-Cyanopyridine-3-carboxylate

  • MDL number MFCD00128863
  • Cas Number : 75358-89-3
  • Molecular Weight : 162.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY008638&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY008638.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart