AC5372910414 Accela ChemBio Inc.

Ethyl (1R,2S)-2-(Boc-amino)cyclopentanecarboxylate

  • MDL number MFCD16883074
  • Cas Number : 1140972-29-7
  • Molecular Weight : 257.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY008647&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY008647.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart