AC5372910420 Accela ChemBio Inc.

(1S,2S,5R)-3-Boc-2-[(tert-butyldimethylsilyloxy)methyl]-4-oxo-3-azabicyclo[3.1.0]hexane

  • MDL number MFCD17171373
  • Cas Number : 220623-07-4
  • Molecular Weight : 341.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY008679&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY008679.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart