AC5372910422 Accela ChemBio Inc.

3-Acetoxy-2-phenylpropanoic Acid

  • MDL number MFCD09750984
  • Cas Number : 14510-36-2
  • Molecular Weight : 208.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY008685&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY008685.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart