AC5372910423 Accela ChemBio Inc.

(8R,9S,10R,13S,14S)-10,13-Dimethyl-7,8,9,11,12,13,15,16-octahydro-10H-cyclopenta[a]phenanthrene-3,6,17(14H)-trione

  • MDL number MFCD19442767
  • Cas Number : 72648-46-5
  • Molecular Weight : 298.37618

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY008686&attname=GoodsCode

Safety Information

hcodes No data known
MSDS http://pdf.accelachem.com/cn/msds/SY008686.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart